accessibility menu, dialog, popup

UserName

H3COClN

CAS RN 17228-64-7

IUPAC Name: 2-chloro-6-methoxypyridine
Synonyms: pyridine, 2-chloro-6-methoxy-
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: VAVGOGHLNAJECD-UHFFFAOYSA-N
SMILES: COC1=CC=CC(Cl)=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)