missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-hydroxynicotinic Acid 97.0+%, TCI America™

Catalog No. C23985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23985G 5 g
C23981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23985G Supplier TCI America Supplier No. C23985G

Chemical Identifiers

CAS 38025-90-0
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.552
MDL Number MFCD09757508
InChI Key BSCFFOOWFVZSSK-UHFFFAOYSA-N
Synonym 2-Chloro-6-hydroxy-3-pyridinecarboxylic Acid
PubChem CID 29919580
IUPAC Name 2-chloro-6-oxo-1H-pyridine-3-carboxylic acid
SMILES C1=CC(=O)NC(=C1C(=O)O)Cl

Specifications

Melting Point 302°C
Color Red-White
Quantity 5 g
Formula Weight 173.55
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-hydroxynicotinic Acid