accessibility menu, dialog, popup

UserName

CAS RN 38025-90-0

OHNOOHCl

CAS RN 38025-90-0

IUPAC Name: 2-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid
Synonyms: 2-chloro-6-oxo-1H-pyridine-3-carboxylic acid
Molecular Weight (g/mol): 173.55
Molecular Formula: C6H4ClNO3
InChi Key: BSCFFOOWFVZSSK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)NC(=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results