missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzyl Alcohol 97.0+%, TCI America™

Catalog No. C163825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C163825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C163825G Supplier TCI America Supplier No. C163825G

Chemical Identifiers

CAS 56456-50-9
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.572
MDL Number MFCD00004608
InChI Key ZZFCUESFXBCRSC-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzyl alcohol, 2-chloro-6-fluorophenyl methanol, 2-chloro-6-fluorobenzylalcohol, 2-fluoro-6-chlorobenzyl alcohol, unii-5k0sc480hm, benzenemethanol, 2-chloro-6-fluoro, 6-fluoro-2-chlorobenzyl alcohol, 2-chloro-6-fluorobenzylic alcohol, pubchem3427, acmc-1axh3
PubChem CID 91868
IUPAC Name (2-chloro-6-fluorophenyl)methanol
SMILES C1=CC(=C(C(=C1)Cl)CO)F

Specifications

Melting Point 43°C
Color White
Quantity 25 g
Formula Weight 160.57
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-fluorobenzyl Alcohol