Learn More
2-Chloro-6-fluorobenzyl alcohol, 97%
CAS: 56456-50-9 | C7H6ClFO | 160.572 g/mol
$55.78 - $152.86
Chemical Identifiers
| CAS | 56456-50-9 |
|---|---|
| Molecular Formula | C7H6ClFO |
| Molecular Weight (g/mol) | 160.572 |
| MDL Number | MFCD00004608 |
| InChI Key | ZZFCUESFXBCRSC-UHFFFAOYSA-N |
| Synonym | 2-chloro-6-fluorobenzyl alcohol, 2-chloro-6-fluorophenyl methanol, 2-chloro-6-fluorobenzylalcohol, 2-fluoro-6-chlorobenzyl alcohol, unii-5k0sc480hm, benzenemethanol, 2-chloro-6-fluoro, 6-fluoro-2-chlorobenzyl alcohol, 2-chloro-6-fluorobenzylic alcohol, pubchem3427, acmc-1axh3 |
| PubChem CID | 91868 |
| IUPAC Name | (2-chloro-6-fluorophenyl)methanol |
| SMILES | C1=CC(=C(C(=C1)Cl)CO)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1197806
|
Thermo Scientific Chemicals
A1197806 |
5 g |
N/A
|
|
|||||
|
AAA1197814
|
Thermo Scientific Chemicals
A1197814 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56456-50-9 | |
| 160.572 | |
| ZZFCUESFXBCRSC-UHFFFAOYSA-N | |
| 91868 | |
| C1=CC(=C(C(=C1)Cl)CO)F |
| C7H6ClFO | |
| MFCD00004608 | |
| 2-chloro-6-fluorobenzyl alcohol, 2-chloro-6-fluorophenyl methanol, 2-chloro-6-fluorobenzylalcohol, 2-fluoro-6-chlorobenzyl alcohol, unii-5k0sc480hm, benzenemethanol, 2-chloro-6-fluoro, 6-fluoro-2-chlorobenzyl alcohol, 2-chloro-6-fluorobenzylic alcohol, pubchem3427, acmc-1axh3 | |
| (2-chloro-6-fluorophenyl)methanol |
Specifications
| 56456-50-9 | |
| >110°C (230°F) | |
| MFCD00004608 | |
| 1938353 | |
| ZZFCUESFXBCRSC-UHFFFAOYSA-N | |
| (2-chloro-6-fluorophenyl)methanol | |
| 91868 | |
| 97% |
| 42°C to 44°C | |
| C7H6ClFO | |
| 5 g | |
| 2-chloro-6-fluorobenzyl alcohol, 2-chloro-6-fluorophenyl methanol, 2-chloro-6-fluorobenzylalcohol, 2-fluoro-6-chlorobenzyl alcohol, unii-5k0sc480hm, benzenemethanol, 2-chloro-6-fluoro, 6-fluoro-2-chlorobenzyl alcohol, 2-chloro-6-fluorobenzylic alcohol, pubchem3427, acmc-1axh3 | |
| C1=CC(=C(C(=C1)Cl)CO)F | |
| 160.572 | |
| 160.58 | |
| 2-Chloro-6-fluorobenzyl alcohol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 260-192-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only