missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzyl alcohol, 97%

Catalog No. AAA1197814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1197814 25 g
AAA1197806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1197814 Supplier Thermo Scientific Chemicals Supplier No. A1197814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56456-50-9 | C7H6ClFO | 160.572 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56456-50-9
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.572
MDL Number MFCD00004608
InChI Key ZZFCUESFXBCRSC-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzyl alcohol, 2-chloro-6-fluorophenyl methanol, 2-chloro-6-fluorobenzylalcohol, 2-fluoro-6-chlorobenzyl alcohol, unii-5k0sc480hm, benzenemethanol, 2-chloro-6-fluoro, 6-fluoro-2-chlorobenzyl alcohol, 2-chloro-6-fluorobenzylic alcohol, pubchem3427, acmc-1axh3
PubChem CID 91868
IUPAC Name (2-chloro-6-fluorophenyl)methanol
SMILES C1=CC(=C(C(=C1)Cl)CO)F

Specifications

Melting Point 42°C to 44°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1938353
Formula Weight 160.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 260-192-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only