missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzaldoxime, 97%

Catalog No. AAL1471903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1471903 1 g
AAL1471906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1471903 Supplier Thermo Scientific Chemicals Supplier No. L1471903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 443-33-4 | C7H5ClFNO | 173.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 443-33-4
Molecular Formula C7H5ClFNO
Molecular Weight (g/mol) 173.57
MDL Number MFCD00837704
InChI Key OBJHLLOVMKKXDI-ONNFQVAWSA-N
Synonym 2-chloro-6-fluorobenzaldehyde oxime, 2-chloro-6-fluorobenzaldoxime, 6-chloro-2-fluorophenyl hydroxyimino methane, pubchem2883, e-n-2-chloro-6-fluorophenyl methylidene hydroxylamine, 1e-2-chloro-6-fluorobenzaldehyde oxime, e-1-2-chloro-6-fluorophenyl-n-hydroxymethanimine, ne-n-2-chloro-6-fluorophenyl methylidene hydroxylamine, ne-n-2-chloranyl-6-fluoranyl-phenyl methylidene hydroxylamine
PubChem CID 7329318
SMILES O\N=C\C1=C(F)C=CC=C1Cl

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Beilstein 3247886
IUPAC Name (E)-N-[(2-chloro-6-fluorophenyl)methylidene]hydroxylamine
Formula Weight 173.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluorobenzaldoxime
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-135-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only