missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-ethoxypyridine 97.0+%, TCI America™

Catalog No. C114325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C114325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C114325G Supplier TCI America Supplier No. C114325G

Chemical Identifiers

CAS 42144-78-5
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00023465
InChI Key AMSLPXHLKHZWBJ-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-6-ethoxy, acmc-209jns, 6-chloro-2-ethoxypyridine, 2-chloro-6-ethoxypyridine, 2-chloro-6-ethoxy-pyridine, pyridine,2-chloro-6-ethoxy, 2-chloranyl-6-ethoxy-pyridine
PubChem CID 98563
IUPAC Name 2-chloro-6-ethoxypyridine
SMILES CCOC1=NC(=CC=C1)Cl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 157.60
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-ethoxypyridine