CAS RN 42144-78-5
IUPAC Name:
2-chloro-6-ethoxypyridine
Synonyms:
2-chloro-6-ethoxypyridine
Molecular Weight (g/mol):
157.6
Molecular Formula:
C7H8ClNO
InChi Key:
AMSLPXHLKHZWBJ-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC(Cl)=N1
1
–
1
of
1
results
1
2-Chloro-6-ethoxypyridine 97.0+%, TCI America™
CAS: 42144-78-5 Molecular Formula: C7H8ClNO Molecular Weight (g/mol): 157.597 MDL Number: MFCD00023465 InChI Key: AMSLPXHLKHZWBJ-UHFFFAOYSA-N Synonym: pyridine, 2-chloro-6-ethoxy,acmc-209jns,6-chloro-2-ethoxypyridine,2-chloro-6-ethoxypyridine,2-chloro-6-ethoxy-pyridine,pyridine,2-chloro-6-ethoxy,2-chloranyl-6-ethoxy-pyridine PubChem CID: 98563 IUPAC Name: 2-chloro-6-ethoxypyridine SMILES: CCOC1=NC(=CC=C1)Cl