missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$74.56 - $74.56
Specifications
| CAS | 182344-18-9 |
|---|---|
| Melting Point | 108°C |
| Color | White |
| Molecular Formula | C7H5BClF3O2 |
| MDL Number | MFCD00797335 |
Specifications
| 182344-18-9 | |
| White | |
| MFCD00797335 | |
| B(C1=C(C=CC(=C1)C(F)(F)F)Cl)(O)O | |
| 224.37 | |
| 224.37 |
| 108°C | |
| C7H5BClF3O2 | |
| YVMXEHZEYONARR-UHFFFAOYSA-N | |
| [2-chloro-5-(trifluoromethyl)phenyl]boronic acid | |
| 2782670 | |
| Crystalline Powder |