missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C25911G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25911G Supplier TCI America Supplier No. C25911G

Specifications

Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 182344-18-9
Melting Point 108°C
Color White
Molecular Formula C7H5BClF3O2
Quantity 1 g
MDL Number MFCD00797335
Synonym 2-chloro-5-trifluoromethyl phenylboronic acid, 2-chloro-5-trifluoromethyl benzeneboronic acid, 2-chloro-5-trifluoromethylphenylboronic acid, 2-chloro-5-trifluoromethyl phenyl boronic acid, 2-chloro-5-trifluoromethyl phenylboronicacid, boronic acid, 2-chloro-5-trifluoromethyl phenyl, pubchem1800, acmc-1c23a, ksc489k6p
InChI Key YVMXEHZEYONARR-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)C(F)(F)F)Cl)(O)O
IUPAC Name [2-chloro-5-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 224.37
PubChem CID 2782670
Formula Weight 224.37
Physical Form Crystalline Powder
Show More Show Less