missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)phenyl Isocyanate 98.0+%, TCI America™

Catalog No. C22585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22585G Supplier TCI America Supplier No. C22585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50528-86-4
Molecular Formula C8H3ClF3NO
Molecular Weight (g/mol) 221.563
MDL Number MFCD00037029
InChI Key WEPYOPYMWSHRIW-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl phenyl isocyanate, 1-chloro-2-isocyanato-4-trifluoromethyl benzene, isocyanic acid 2-chloro-5-trifluoromethyl phenyl ester, 2-chloro-5-trifluoromethylphenyl isocyanate, 2-chloro-5-trifluoromethyl phenylisocyanate, 2-chloro-5-trifluoromethyl benzenisocyanate, pubchem5026, acmc-1aq1j, timtec-bb sbb006656, 1-mercapto-2-methyl-propan-2-ol
PubChem CID 2733263
IUPAC Name 1-chloro-2-isocyanato-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)N=C=O)Cl

Specifications

Color Yellow
Boiling Point 64°C
Quantity 5 g
UN Number 2206
Formula Weight 221.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenyl Isocyanate
TSCA No
Recommended Storage Refrigerator