missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)phenyl isocyanate, 97%

Catalog No. AAL0989906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0989906 5 g
AAL0989903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0989906 Supplier Thermo Scientific Chemicals Supplier No. L0989906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 50528-86-4 | C8H3ClF3NO | 221.563 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50528-86-4
Molecular Formula C8H3ClF3NO
Molecular Weight (g/mol) 221.563
MDL Number MFCD00037029
InChI Key WEPYOPYMWSHRIW-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl phenyl isocyanate, 1-chloro-2-isocyanato-4-trifluoromethyl benzene, isocyanic acid 2-chloro-5-trifluoromethyl phenyl ester, 2-chloro-5-trifluoromethylphenyl isocyanate, 2-chloro-5-trifluoromethyl phenylisocyanate, 2-chloro-5-trifluoromethyl benzenisocyanate, pubchem5026, acmc-1aq1j, timtec-bb sbb006656, 1-mercapto-2-methyl-propan-2-ol
PubChem CID 2733263
IUPAC Name 1-chloro-2-isocyanato-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)N=C=O)Cl

Specifications

Density 1.48
Boiling Point 52°C to 53°C (3 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.489
Quantity 5 g
UN Number UN2206
Beilstein 2112804
Sensitivity Moisture sensitive
Formula Weight 221.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenyl isocyanate
Show More Show Less
Hazard Category H227-H302-H315-H319-H331-H334-H335
Hazard Statement GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only