missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. p-7125255
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20505G 5 g
C205025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20505G Supplier TCI America Supplier No. C20505G
Only null left

Chemical Identifiers

CAS 328-87-0
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.564
InChI Key LCISFYAQKHOWBP-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl benzonitrile, benzonitrile, 2-chloro-5-trifluoromethyl, 4-chloro-3-cyanobenzotrifluoride, 4-chloro-3-cyanobenzo trifluoride, 4-chloro-3-cyanbenzotrifluoride, 2-chloro-5-trifluoromethyl benzenecarbonitrile, ncr bg exfff, pubchem4749, acmc-1cq7c, ksc494q7d
PubChem CID 67604
IUPAC Name 2-chloro-5-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C(F)(F)F)C#N)Cl

Specifications

Melting Point 39°C
Boiling Point 212°C
Quantity 5 g
UN Number 3439
Formula Weight 205.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzonitrile