missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 96%

Catalog No. p-7048021
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2010403 1 g
AAL2010406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2010403 Supplier Thermo Scientific Chemicals Supplier No. L2010403

CAS: 182344-18-9 | C7H5BClF3O2 | 224.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 182344-18-9
Molecular Formula C7H5BClF3O2
Molecular Weight (g/mol) 224.37
MDL Number MFCD00797335
InChI Key YVMXEHZEYONARR-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl phenylboronic acid, 2-chloro-5-trifluoromethyl benzeneboronic acid, 2-chloro-5-trifluoromethylphenylboronic acid, 2-chloro-5-trifluoromethyl phenyl boronic acid, 2-chloro-5-trifluoromethyl phenylboronicacid, boronic acid, 2-chloro-5-trifluoromethyl phenyl, pubchem1800, acmc-1c23a, ksc489k6p
PubChem CID 2782670
IUPAC Name [2-chloro-5-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(F)(F)F)Cl)(O)O

Specifications

Quantity 1 g
Formula Weight 224.38
Percent Purity 96%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only