missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)benzaldehyde 95.0+%, TCI America™

Catalog No. C20475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20475G 5 g
C204725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20475G Supplier TCI America Supplier No. C20475G
Only null left

Chemical Identifiers

CAS 82386-89-8
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.564
MDL Number MFCD00134430
InChI Key OZZOJJJYKYKBNH-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl benzaldehyde, benzaldehyde, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-benzaldehyde, pubchem2881, acmc-209pop, ksc495k9f, timtec-bb sbb003466, 4-chloro-3-formylbenzotrifluoride
PubChem CID 688181
IUPAC Name 2-chloro-5-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1C(F)(F)F)C=O)Cl

Specifications

Color Yellow
Boiling Point 97°C
Quantity 5 g
Formula Weight 208.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)benzaldehyde