missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloro-5'-(trifluoromethyl)acetophenone 97.0+%, TCI America™

Catalog No. C33361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33361G 1 g
C33365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33361G Supplier TCI America Supplier No. C33361G

Chemical Identifiers

CAS 71648-45-8
Molecular Formula C9H6ClF3O
Molecular Weight (g/mol) 222.591
MDL Number MFCD00045092
InChI Key YRGBMTWHOFQSDJ-UHFFFAOYSA-N
Synonym 2'-chloro-5'-trifluoromethyl acetophenone, 1-2-chloro-5-trifluoromethyl phenyl ethanone, 1-2-chloro-5-trifluoromethyl phenyl ethan-1-one, 2-chloro-5-trifluoromethyl acetophenone, acetophenone, 2-chloro-5-trifluoromethyl, 3-acetyl-4-chlorobenzotrifluoride, ethanone, 1-2-chloro-5-trifluoromethyl phenyl, 1-acetyl-2-chloro-5-trifluoromethyl benzene, pubchem4210
PubChem CID 144502
IUPAC Name 1-[2-chloro-5-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=C(C=CC(=C1)C(F)(F)F)Cl

Specifications

Color Yellow
Boiling Point 205°C
Quantity 1 g
Formula Weight 222.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Chloro-5′-(trifluoromethyl)acetophenone