missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitrotoluene 98.0+%, TCI America™
$96.94 - $736.30
Chemical Identifiers
| CAS | 13290-74-9 |
|---|---|
| Molecular Formula | C7H6ClNO2 |
| Molecular Weight (g/mol) | 171.58 |
| MDL Number | MFCD00041430 |
| InChI Key | BGDCQZFFNFXYQC-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-nitrotoluene, benzene, 1-chloro-2-methyl-4-nitro, 4-chloro-3-methylnitrobenzene, 1-chloro-2-methyl-4-nitro-benzene, 3-nitro-6-chlorotoluene, 2-methyl-4-nitrochlorobenzene, pubchem2137, 2-chloro-5-nitroluene, acmc-209bqf, 5-nitro-2-chlorotoluene |
| PubChem CID | 83315 |
| IUPAC Name | 1-chloro-2-methyl-4-nitrobenzene |
| SMILES | CC1=C(C=CC(=C1)[N+](=O)[O-])Cl |
Chemical Identifiers
| 13290-74-9 | |
| 171.58 | |
| BGDCQZFFNFXYQC-UHFFFAOYSA-N | |
| 83315 | |
| CC1=C(C=CC(=C1)[N+](=O)[O-])Cl |
| C7H6ClNO2 | |
| MFCD00041430 | |
| 2-chloro-5-nitrotoluene, benzene, 1-chloro-2-methyl-4-nitro, 4-chloro-3-methylnitrobenzene, 1-chloro-2-methyl-4-nitro-benzene, 3-nitro-6-chlorotoluene, 2-methyl-4-nitrochlorobenzene, pubchem2137, 2-chloro-5-nitroluene, acmc-209bqf, 5-nitro-2-chlorotoluene | |
| 1-chloro-2-methyl-4-nitrobenzene |
Specifications
| 13290-74-9 | |
| Yellow | |
| C7H6ClNO2 | |
| 25 g | |
| 2-chloro-5-nitrotoluene, benzene, 1-chloro-2-methyl-4-nitro, 4-chloro-3-methylnitrobenzene, 1-chloro-2-methyl-4-nitro-benzene, 3-nitro-6-chlorotoluene, 2-methyl-4-nitrochlorobenzene, pubchem2137, 2-chloro-5-nitroluene, acmc-209bqf, 5-nitro-2-chlorotoluene | |
| CC1=C(C=CC(=C1)[N+](=O)[O-])Cl | |
| 171.58 | |
| 171.58 | |
| Crystalline Powder |
| 44°C | |
| 96°C | |
| MFCD00041430 | |
| 3457 | |
| BGDCQZFFNFXYQC-UHFFFAOYSA-N | |
| 1-chloro-2-methyl-4-nitrobenzene | |
| 83315 | |
| ≥98.0% (GC) | |
| 2-Chloro-5-nitrotoluene |
Safety and Handling
EINECSNumber : (3)-0449
TSCA : Yes