missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitropyrimidine 98.0+%, TCI America™
$121.16 - $594.63
Chemical Identifiers
| CAS | 10320-42-0 |
|---|---|
| Molecular Formula | C4H2ClN3O2 |
| Molecular Weight (g/mol) | 159.529 |
| MDL Number | MFCD04117995 |
| InChI Key | OFCBNMYNAHUDGE-UHFFFAOYSA-N |
| PubChem CID | 82544 |
| IUPAC Name | 2-chloro-5-nitropyrimidine |
| SMILES | C1=C(C=NC(=N1)Cl)[N+](=O)[O-] |
Chemical Identifiers
| 10320-42-0 | |
| 159.529 | |
| OFCBNMYNAHUDGE-UHFFFAOYSA-N | |
| 2-chloro-5-nitropyrimidine |
| C4H2ClN3O2 | |
| MFCD04117995 | |
| 82544 | |
| C1=C(C=NC(=N1)Cl)[N+](=O)[O-] |
Specifications
| 10320-42-0 | |
| Yellow | |
| MFCD04117995 | |
| OFCBNMYNAHUDGE-UHFFFAOYSA-N | |
| 2-chloro-5-nitropyrimidine | |
| 82544 | |
| ≥98.0% (GC) | |
| 2-Chloro-5-nitropyrimidine |
| 109°C | |
| C4H2ClN3O2 | |
| 1 g | |
| C1=C(C=NC(=N1)Cl)[N+](=O)[O-] | |
| 159.529 | |
| 159.53 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer