missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrophenol 98.0+%, TCI America™

Catalog No. p-7125272
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20775G 5 g
C207725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20775G Supplier TCI America Supplier No. C20775G

Chemical Identifiers

CAS 619-10-3
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD01571825
InChI Key BUMGQSCPTLELLS-UHFFFAOYSA-N
Synonym phenol, 2-chloro-5-nitro, 6-chloro-3-nitrophenol, 2-chloro-5-nitro-phenol, pubchem1475, acmc-209mxh, 2-chloro-5-nitrofenol, 5-nitro-2-chlorophenol, 2-chloro-5-nitro phenol, ksc494c3h, 4-chloro-3-hydroxynitrobenzene
PubChem CID 69264
IUPAC Name 2-chloro-5-nitrophenol
SMILES OC1=CC(=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 173.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrophenol