missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrophenol, 98%

Catalog No. p-7077771
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459040050 5 g
AC459040250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459040050 Supplier Thermo Scientific Chemicals Supplier No. 459040050
Only null left

CAS: 619-10-3 | C6H4ClNO3 | 173.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-10-3
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD01571825
InChI Key BUMGQSCPTLELLS-UHFFFAOYSA-N
Synonym phenol, 2-chloro-5-nitro, 6-chloro-3-nitrophenol, 2-chloro-5-nitro-phenol, pubchem1475, acmc-209mxh, 2-chloro-5-nitrofenol, 5-nitro-2-chlorophenol, 2-chloro-5-nitro phenol, ksc494c3h, 4-chloro-3-hydroxynitrobenzene
PubChem CID 69264
IUPAC Name 2-chloro-5-nitrophenol
SMILES OC1=CC(=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 119.0°C to 121.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 173.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-nitrophenol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 210-579-3

RUO – Research Use Only