missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrophenol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 459040050

 View more versions of this product

Catalog No. AC459040050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Chloro-5-nitrophenol
Authentic
C6H4ClNO3
5g
BUMGQSCPTLELLS-UHFFFAOYSA-N
2-chloro-5-nitrophenol
69264
98%
619-10-3
97.5% min. (HPLC)
MFCD01571825
phenol, 2-chloro-5-nitro, 6-chloro-3-nitrophenol, 2-chloro-5-nitro-phenol, pubchem1475, acmc-209mxh, 2-chloro-5-nitrofenol, 5-nitro-2-chlorophenol, 2-chloro-5-nitro phenol, ksc494c3h, 4-chloro-3-hydroxynitrobenzene
OC1=CC(=CC=C1Cl)[N+]([O-])=O
173.55
173.56

Chemical Identifiers

619-10-3
173.55
BUMGQSCPTLELLS-UHFFFAOYSA-N
69264
OC1=CC(=CC=C1Cl)[N+]([O-])=O
C6H4ClNO3
MFCD01571825
phenol, 2-chloro-5-nitro, 6-chloro-3-nitrophenol, 2-chloro-5-nitro-phenol, pubchem1475, acmc-209mxh, 2-chloro-5-nitrofenol, 5-nitro-2-chlorophenol, 2-chloro-5-nitro phenol, ksc494c3h, 4-chloro-3-hydroxynitrobenzene
2-chloro-5-nitrophenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 210-579-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only