missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzyl Alcohol 98.0+%, TCI America™

Catalog No. C12145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12145G Supplier TCI America Supplier No. C12145G

Chemical Identifiers

CAS 80866-80-4
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00007297
InChI Key NNLQYDLTFRXAKD-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrophenyl methanol, 2-chloro-5-nitrobenzyl alcohol, 2-chloro-5-nitrobenzylalcohol, benzenemethanol, 2-chloro-5-nitro, acmc-1bkf2, nnlqydltfrxakd-uhfffaoysa, benzenemethanol,2-chloro-5-nitro, 2-chloro-5-nitro-phenyl-methanol, 2-chloro-5-nitrophenyl methanol #, 2-chloro-5-nitrophenyl methan-1-ol
PubChem CID 555722
IUPAC Name (2-chloro-5-nitrophenyl)methanol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])CO)Cl

Specifications

Melting Point 77°C
Quantity 5 g
Formula Weight 187.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzyl Alcohol