missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzyl alcohol, 98%

Catalog No. AAA1420522
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1420522 100 g
AAA1420506 5 g
AAA1420514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1420522 Supplier Thermo Scientific Chemicals Supplier No. A1420522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80866-80-4 | C7H6ClNO3 | 187.579 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80866-80-4
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00007297
InChI Key NNLQYDLTFRXAKD-UHFFFAOYSA-N
PubChem CID 555722
IUPAC Name (2-chloro-5-nitrophenyl)methanol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])CO)Cl

Specifications

Melting Point 75°C to 79°C
Quantity 100 g
Beilstein 3248759
Synonym 2-chloro-5-nitrophenyl methanol, 2-chloro-5-nitrobenzyl alcohol, 2-chloro-5-nitrobenzylalcohol, benzenemethanol, 2-chloro-5-nitro, acmc-1bkf2, nnlqydltfrxakd-uhfffaoysa, benzenemethanol,2-chloro-5-nitro, 2-chloro-5-nitro-phenyl-methanol, 2-chloro-5-nitrophenyl methanol #, 2-chloro-5-nitrophenyl methan-1-ol
Formula Weight 187.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-nitrobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 279-584-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only