missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzotrifluoride 97.0+%, TCI America™

Catalog No. C092325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C092325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C092325G Supplier TCI America Supplier No. C092325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 777-37-7
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.55
MDL Number MFCD00007296
InChI Key HQROXDLWVGFPDE-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzotrifluoride, 1-chloro-4-nitro-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl nitrobenzene, benzene, 1-chloro-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-chloronitrobenzene, 2-trifluoromethyl-4-nitrochlorobenzene, 2-chloro-5-nitro-a,a,a-trifluorotoluene, 4-nitro-2-trifluoromethyl chlorobenzene, 1-chloro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-2-chlorobenzotrifluoride
PubChem CID 61213
IUPAC Name 1-chloro-4-nitro-2-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 232°C
Quantity 25 g
UN Number 2811
Formula Weight 225.55
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-5-nitrobenzotrifluoride