missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. C20265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20265G 5 g
C202625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20265G Supplier TCI America Supplier No. C20265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16588-02-6
Molecular Formula C7H3ClN2O2
Molecular Weight (g/mol) 182.563
MDL Number MFCD00007292
InChI Key ZGILLTVEEBNDOB-UHFFFAOYSA-N
Synonym benzonitrile, 2-chloro-5-nitro, 2-chlor-5-nitrobenzonitril german, 5-nitro-2-chlorobenzonitrile, chembl52706, 2-chloro-5-nitro-benzonitrile, 2-chloro-5-nitrobenzenecarbonitrile, 4-chloro-3-cyano-1-nitrobenzene, 2-chlor-5-nitrobenzonitril, pubchem4740, 2-chloro-5-nitrobenzonitril
PubChem CID 85504
IUPAC Name 2-chloro-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)Cl

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 182.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzonitrile
EINECSNumber (3)-1781
RTECSNumber DI3040000
TSCA No