missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitrobenzoic Acid 98.0+%, TCI America™
$50.05 - $327.28
Chemical Identifiers
| CAS | 2516-96-3 |
|---|---|
| Molecular Formula | C7H4ClNO4 |
| Molecular Weight (g/mol) | 201.562 |
| MDL Number | MFCD00007294 |
| InChI Key | QUEKGYQTRJVEQC-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-nitrobenzoic acid, benzoic acid, 2-chloro-5-nitro, 6-chloro-3-nitrobenzoic acid, 2-chloro-5-nitro-benzoic acid, 2-chloro-5-nitrobenzoicacid, mesalamine impurity m, pubchem13682, acmc-1cb0i, 2-chloro-5-nitrobenzic acid, 5-nitro-2-chlorobenzoic acid |
| PubChem CID | 17287 |
| SMILES | C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Cl |
Chemical Identifiers
| 2516-96-3 | |
| 201.562 | |
| QUEKGYQTRJVEQC-UHFFFAOYSA-N | |
| 17287 |
| C7H4ClNO4 | |
| MFCD00007294 | |
| 2-chloro-5-nitrobenzoic acid, benzoic acid, 2-chloro-5-nitro, 6-chloro-3-nitrobenzoic acid, 2-chloro-5-nitro-benzoic acid, 2-chloro-5-nitrobenzoicacid, mesalamine impurity m, pubchem13682, acmc-1cb0i, 2-chloro-5-nitrobenzic acid, 5-nitro-2-chlorobenzoic acid | |
| C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Cl |
Specifications
| 2516-96-3 | |
| White-Yellow | |
| MFCD00007294 | |
| 2-chloro-5-nitrobenzoic acid, benzoic acid, 2-chloro-5-nitro, 6-chloro-3-nitrobenzoic acid, 2-chloro-5-nitro-benzoic acid, 2-chloro-5-nitrobenzoicacid, mesalamine impurity m, pubchem13682, acmc-1cb0i, 2-chloro-5-nitrobenzic acid, 5-nitro-2-chlorobenzoic acid | |
| C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Cl | |
| 17287 | |
| ≥98.0% (GC,T) | |
| 2-Chloro-5-nitrobenzoic Acid |
| 165°C | |
| C7H4ClNO4 | |
| 25 g | |
| QUEKGYQTRJVEQC-UHFFFAOYSA-N | |
| 201.562 | |
| 201.56 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1502
TSCA : Yes