missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitroanisole 98.0+%, TCI America™

Catalog No. c17735g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17735G 5 g
C177325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17735G Supplier TCI America Supplier No. C17735G
Only null left

Chemical Identifiers

CAS 1009-36-5
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
MDL Number MFCD00079739
InChI Key JXIJUAWSDBACEB-UHFFFAOYSA-N
Synonym 2-Methoxy-4-nitrochlorobenzene
PubChem CID 70519
IUPAC Name 1-chloro-2-methoxy-4-nitrobenzene
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])Cl

Specifications

Melting Point 83°C
Quantity 5 g
Formula Weight 187.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitroanisole
EINECSNumber (3)-0910&(3)-0911
TSCA Yes