missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitroanisole 98.0+%, TCI America™
Supplier: TCI America C177325G
Specifications
| 2-Chloro-5-nitroanisole | |
| 83°C | |
| MFCD00079739 | |
| 2-Methoxy-4-nitrochlorobenzene | |
| COC1=C(C=CC(=C1)[N+](=O)[O-])Cl | |
| 187.579 | |
| 187.58 | |
| Crystalline Powder |
| 1009-36-5 | |
| C7H6ClNO3 | |
| 25 g | |
| JXIJUAWSDBACEB-UHFFFAOYSA-N | |
| 1-chloro-2-methoxy-4-nitrobenzene | |
| 70519 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1009-36-5 | |
| 187.579 | |
| JXIJUAWSDBACEB-UHFFFAOYSA-N | |
| 70519 | |
| COC1=C(C=CC(=C1)[N+](=O)[O-])Cl |
| C7H6ClNO3 | |
| MFCD00079739 | |
| 2-Methoxy-4-nitrochlorobenzene | |
| 1-chloro-2-methoxy-4-nitrobenzene |
Safety and Handling
EINECSNumber : (3)-0910&(3)-0911
TSCA : Yes