missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-methylphenol, 99%
Description
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
Solubility
Slightly Soluble (1.2 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
2-Chloro-5-methylphenol, is used as a pharmaceutical intermediate. It is also used to make other chemicals.
Solubility
Slightly Soluble (1.2 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 44°C to 48°C |
| Density | 1.215 |
| Boiling Point | 195°C to 197°C |
| Flash Point | 81°C (177°F) |
| Quantity | 25 g |
| UN Number | UN3437 |
| Beilstein | 2041487 |
| Solubility Information | Slightly Soluble (1.2g/L) (25°C). |
| Formula Weight | 142.59 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Hazard Category | H302+H312-H317-H318-H335 |
| Hazard Statement | GHS H Statement H301-H311-H331-H318-H315-H335 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes serious eye damage. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID |
| EINECSNumber | 210-444-9 |
| RTECSNumber | SK4930000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only