missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. C17801G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17801G Supplier TCI America Supplier No. C17801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6342-60-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00045798
InChI Key LEBWXJZAWTVKFL-UHFFFAOYSA-N
Synonym 2-chloro-5-methylbenzole acid, 6-chloro-m-toluic acid, benzoic acid, 2-chloro-5-methyl, pubchem12501, 3-carboxy-4-chlorotoluene, acmc-1b72o, ksc497q1d, 2-chloro-5-methylbenozic acid, 2-chloro-5-methyl-benzoic acid, lebwxjzawtvkfl-uhfffaoysa
PubChem CID 240430
IUPAC Name 2-chloro-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)Cl)C(=O)O

Specifications

Melting Point 150°C
Quantity 1 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-methylbenzoic Acid