missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-methylbenzoic acid, 98%

Catalog No. p-4258898
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC381970010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC381970010 Supplier Thermo Scientific Chemicals Supplier No. 381970010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6342-60-5 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6342-60-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
InChI Key LEBWXJZAWTVKFL-UHFFFAOYSA-N
Synonym 2-chloro-5-methylbenzole acid, 6-chloro-m-toluic acid, benzoic acid, 2-chloro-5-methyl, pubchem12501, 3-carboxy-4-chlorotoluene, acmc-1b72o, ksc497q1d, 2-chloro-5-methylbenozic acid, 2-chloro-5-methyl-benzoic acid, lebwxjzawtvkfl-uhfffaoysa
PubChem CID 240430
IUPAC Name 2-chloro-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)Cl)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 148.0°C to 151.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(Cl)COOH
Quantity 1 g
Formula Weight 170.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-chloro-5-methylbenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only