missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxypyrimidine 98.0+%, TCI America™

Catalog No. C26935G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26935G 5 g
C26931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26935G Supplier TCI America Supplier No. C26935G

Chemical Identifiers

CAS 22536-65-8
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.558
MDL Number MFCD08702770
InChI Key RSUBGBZOMBTDTI-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-pyrimidine, pubchem13187, acmc-20a0mo, 2-chlor-5-methoxy-pyrimidine, ksc494k1t, 2-chloranyl-5-methoxy-pyrimidine, 2-chloro-5-methoxypyrimidine, pyrimidine, 2-chloro-5-methoxy-8ci,9ci
PubChem CID 589058
IUPAC Name 2-chloro-5-methoxypyrimidine
SMILES COC1=CN=C(N=C1)Cl

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 144.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-methoxypyrimidine