accessibility menu, dialog, popup

UserName

CAS RN 22536-65-8

IUPAC Name: 2-chloro-5-methoxypyrimidine
Synonyms: 2-chloro-5-methoxypyrimidine
Molecular Weight (g/mol): 144.56
Molecular Formula: C5H5ClN2O
InChi Key: RSUBGBZOMBTDTI-UHFFFAOYSA-N
SMILES: COC1=CN=C(Cl)N=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results