missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-methoxybenzoic Acid 98.0+%, TCI America™
Supplier: TCI America C27551G
Specifications
| 2-Chloro-5-methoxybenzoic Acid | |
| 174°C | |
| MFCD00466261 | |
| 2-chloro-5-methoxy-benzoic acid, 6-chloro-m-anisic acid, benzoic acid, 2-chloro-5-methoxy, 2-chloro-5-methoxybenzoicacid, pubchem4563, zlchem 1016, acmc-20a0mp, 3-carboxy-4-chloroanisole, 2-chloro-5-methoxybenzoic acid | |
| COC1=CC=C(Cl)C(=C1)C(O)=O | |
| 186.59 | |
| 186.59 | |
| Crystalline Powder |
| 6280-89-3 | |
| C8H7ClO3 | |
| 1 g | |
| AQHFCRYZABKUEV-UHFFFAOYSA-N | |
| 2-chloro-5-methoxybenzoic acid | |
| 221503 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 6280-89-3 | |
| 186.59 | |
| AQHFCRYZABKUEV-UHFFFAOYSA-N | |
| 221503 | |
| COC1=CC=C(Cl)C(=C1)C(O)=O |
| C8H7ClO3 | |
| MFCD00466261 | |
| 2-chloro-5-methoxy-benzoic acid, 6-chloro-m-anisic acid, benzoic acid, 2-chloro-5-methoxy, 2-chloro-5-methoxybenzoicacid, pubchem4563, zlchem 1016, acmc-20a0mp, 3-carboxy-4-chloroanisole, 2-chloro-5-methoxybenzoic acid | |
| 2-chloro-5-methoxybenzoic acid |
Safety and Handling
TSCA : No