missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. C2755200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2755200MG 200 mg
C27551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2755200MG Supplier TCI America Supplier No. C2755200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6280-89-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00466261
InChI Key AQHFCRYZABKUEV-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxy-benzoic acid, 6-chloro-m-anisic acid, benzoic acid, 2-chloro-5-methoxy, 2-chloro-5-methoxybenzoicacid, pubchem4563, zlchem 1016, acmc-20a0mp, 3-carboxy-4-chloroanisole, 2-chloro-5-methoxybenzoic acid
PubChem CID 221503
IUPAC Name 2-chloro-5-methoxybenzoic acid
SMILES COC1=CC=C(Cl)C(=C1)C(O)=O

Specifications

Melting Point 174°C
Quantity 200 mg
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-methoxybenzoic Acid