missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodobenzotrifluoride 98.0+%, TCI America™

Catalog No. C21965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21965G 5 g
C219625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. C21965G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C21965G
Only null left

Chemical Identifiers

CAS 260355-20-2
Molecular Formula C7H3ClF3I
Molecular Weight (g/mol) 306.45
MDL Number MFCD09037801
InChI Key MCIFWVOVVPVBRJ-UHFFFAOYSA-N
Synonym 1-Chloro-4-iodo-2-(trifluoromethyl)benzene
PubChem CID 43145392
IUPAC Name 1-chloro-4-iodo-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=C(Cl)C=CC(I)=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 306.45
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Chloro-5-iodobenzotrifluoride