missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodobenzonitrile 98.0+%, TCI America™

Catalog No. C22121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22121G 1 g
C22125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22121G Supplier TCI America Supplier No. C22121G
Only null left

Chemical Identifiers

CAS 289039-29-8
Molecular Formula C7H3ClIN
Molecular Weight (g/mol) 263.46
MDL Number MFCD00672973
InChI Key RXNOBNYCCAEGJD-UHFFFAOYSA-N
PubChem CID 24721573
IUPAC Name 2-chloro-5-iodobenzonitrile
SMILES ClC1=C(C=C(I)C=C1)C#N

Specifications

Melting Point 117°C
Quantity 1 g
UN Number 3439
Formula Weight 263.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-iodobenzonitrile