missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoropyrimidine 98.0+%, TCI America™

Catalog No. C25461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25461G 1 g
C25465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25461G Supplier TCI America Supplier No. C25461G

Chemical Identifiers

CAS 62802-42-0
Molecular Formula C4H2ClFN2
Molecular Weight (g/mol) 132.522
MDL Number MFCD03788197
InChI Key AGYUQBNABXVWMS-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine
PubChem CID 583466
IUPAC Name 2-chloro-5-fluoropyrimidine
SMILES C1=C(C=NC(=N1)Cl)F

Specifications

Color Yellow
Boiling Point 94°C
Quantity 1 g
Formula Weight 132.52
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluoropyrimidine