accessibility menu, dialog, popup

UserName

CAS RN 62802-42-0

IUPAC Name: 2-chloro-5-fluoropyrimidine
Synonyms: 2-chloro-5-fluoropyrimidine
Molecular Weight (g/mol): 132.52
Molecular Formula: C4H2ClFN2
InChi Key: AGYUQBNABXVWMS-UHFFFAOYSA-N
SMILES: FC1=CN=C(Cl)N=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)