Learn More
2-chloro-5-fluoropyrimidine, 98%, Thermo Scientific™
$408.98
Chemical Identifiers
| CAS | 62802-42-0 |
|---|---|
| Molecular Formula | C4H2ClFN2 |
| Molecular Weight (g/mol) | 132.53 |
| MDL Number | MFCD03788197 |
| InChI Key | AGYUQBNABXVWMS-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine |
| PubChem CID | 583466 |
| IUPAC Name | 2-chloro-5-fluoropyrimidine |
| SMILES | C1=C(C=NC(=N1)Cl)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC429812500
|
N/A | 250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC429810010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 62802-42-0 | |
| 132.53 | |
| AGYUQBNABXVWMS-UHFFFAOYSA-N | |
| 583466 | |
| C1=C(C=NC(=N1)Cl)F |
| C4H2ClFN2 | |
| MFCD03788197 | |
| 2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine | |
| 2-chloro-5-fluoropyrimidine |
Specifications
| 62802-42-0 | |
| 100.0 | |
| 65°C | |
| 97.5% min. (GC) | |
| MFCD03788197 | |
| AGYUQBNABXVWMS-UHFFFAOYSA-N | |
| 2-chloro-5-fluoropyrimidine | |
| 583466 | |
| 98% |
| 97.5 | |
| 149°C to 162°C (1013.0 hPa) | |
| Authentic | |
| C4H2ClFN2 | |
| 2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine | |
| C1=C(C=NC(=N1)Cl)F | |
| 132.53 | |
| 132.53 | |
| 2-chloro-5-fluoropyrimidine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air a
GHS Signal Word: Danger
RUO – Research Use Only