missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorobenzylamine 98.0+%, TCI America™

Catalog No. p-7125850
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
C31401G 1 g
C31405G 5 g
2 options

Catalog No. C31401G

Supplier: TCI America C31401G

Only null left

Chemical Identifiers

CAS 202522-23-4
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD06213626
InChI Key IGFBMQSUHFQVFE-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorobenzylamine, 2-chloro-5-fluorobenzyl amine, 2-chloro-5-fluorophenyl methanamine, 2-chloro-5-fluoro-benzylamine, 2-chloro-5-fluorophenyl methylamine, 1-2-chloro-5-fluorophenyl methanamine, pubchem4371, acmc-1cio4, rarechem al bw 2058
PubChem CID 17750669
IUPAC Name (2-chloro-5-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1F)CN)Cl

Specifications

Color Yellow
Boiling Point 72°C
Quantity 1 g
UN Number 2735
Formula Weight 159.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluorobenzylamine