missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorobenzylamine 98.0+%, TCI America™

Catalog No. C31405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31405G 5 g
C31401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31405G Supplier TCI America Supplier No. C31405G

Chemical Identifiers

CAS 202522-23-4
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.588
MDL Number MFCD06213626
InChI Key IGFBMQSUHFQVFE-UHFFFAOYSA-N
Synonym 2-chloro-5-fluorobenzylamine, 2-chloro-5-fluorobenzyl amine, 2-chloro-5-fluorophenyl methanamine, 2-chloro-5-fluoro-benzylamine, 2-chloro-5-fluorophenyl methylamine, 1-2-chloro-5-fluorophenyl methanamine, pubchem4371, acmc-1cio4, rarechem al bw 2058
PubChem CID 17750669
IUPAC Name (2-chloro-5-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1F)CN)Cl

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 g
UN Number 2735
Formula Weight 159.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluorobenzylamine