missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-fluorobenzyl Bromide 96.0+%, TCI America™
$83.89 - $242.33
Chemical Identifiers
| CAS | 81778-09-8 |
|---|---|
| Molecular Formula | C7H5BrClF |
| Molecular Weight (g/mol) | 223.469 |
| MDL Number | MFCD01631419 |
| InChI Key | AUVLFQDKJFSFIX-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-fluorobenzyl bromide, 2-bromomethyl-1-chloro-4-fluorobenzene, 2-chloro-5-fluorobenzylbromide, 1-bromomethyl-2-chloro-5-fluorobenzene, pubchem7498, acmc-1blqd, 2-bromomethyl-1-chloro-4-fluoro-benzene, 2-chloro5-fluorobenzyl bromide, 2-chloro-5-fluoro-benzylbromide, 2-chloro-5-fluoro-benzyl bromide |
| PubChem CID | 2773623 |
| IUPAC Name | 2-(bromomethyl)-1-chloro-4-fluorobenzene |
| SMILES | C1=CC(=C(C=C1F)CBr)Cl |
Chemical Identifiers
| 81778-09-8 | |
| 223.469 | |
| AUVLFQDKJFSFIX-UHFFFAOYSA-N | |
| 2773623 | |
| C1=CC(=C(C=C1F)CBr)Cl |
| C7H5BrClF | |
| MFCD01631419 | |
| 2-chloro-5-fluorobenzyl bromide, 2-bromomethyl-1-chloro-4-fluorobenzene, 2-chloro-5-fluorobenzylbromide, 1-bromomethyl-2-chloro-5-fluorobenzene, pubchem7498, acmc-1blqd, 2-bromomethyl-1-chloro-4-fluoro-benzene, 2-chloro5-fluorobenzyl bromide, 2-chloro-5-fluoro-benzylbromide, 2-chloro-5-fluoro-benzyl bromide | |
| 2-(bromomethyl)-1-chloro-4-fluorobenzene |
Specifications
| 81778-09-8 | |
| 66°C | |
| MFCD01631419 | |
| 2924 | |
| AUVLFQDKJFSFIX-UHFFFAOYSA-N | |
| 2-(bromomethyl)-1-chloro-4-fluorobenzene | |
| 2773623 | |
| ≥96.0% (GC) | |
| 2-Chloro-5-fluorobenzyl Bromide |
| Yellow | |
| C7H5BrClF | |
| 1 g | |
| 2-chloro-5-fluorobenzyl bromide, 2-bromomethyl-1-chloro-4-fluorobenzene, 2-chloro-5-fluorobenzylbromide, 1-bromomethyl-2-chloro-5-fluorobenzene, pubchem7498, acmc-1blqd, 2-bromomethyl-1-chloro-4-fluoro-benzene, 2-chloro5-fluorobenzyl bromide, 2-chloro-5-fluoro-benzylbromide, 2-chloro-5-fluoro-benzyl bromide | |
| C1=CC(=C(C=C1F)CBr)Cl | |
| 223.469 | |
| 223.47 | |
| Liquid |
Safety and Handling
TSCA : No