missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoro-4-pyrimidinone 98.0+%, TCI America™

Catalog No. C33815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33815G 5 g
C33811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33815G Supplier TCI America Supplier No. C33815G
Only null left

Chemical Identifiers

CAS 155-12-4
Molecular Formula C4H2ClFN2O
Molecular Weight (g/mol) 148.521
MDL Number MFCD01346689
InChI Key FHTLMXYURUPWJB-UHFFFAOYSA-N
PubChem CID 255394
IUPAC Name 2-chloro-5-fluoro-1H-pyrimidin-6-one
SMILES C1=C(C(=O)NC(=N1)Cl)F

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 5 g
Formula Weight 148.52
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-fluoro-4-pyrimidinone