missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-cyanopyridine 97.0+%, TCI America™
Supplier: TCI America C205625G
Specifications
| 2-Chloro-5-cyanopyridine | |
| 117°C | |
| C6H3ClN2 | |
| 25 g | |
| 2-chloro-5-cyanopyridine, 6-chloronicotinonitrile, 2-chloropyridine-5-carbonitrile, 6-chloro-3-pyridinecarbonitrile, 6-chloronicotinitrile, 6-chloronicotinontrile, 6-chloro-nicotinonitrile, 2-chloro-5-cyano-pyridine, 3-pyridinecarbonitrile, 6-chloro, 6-chloro nicotinonitrile | |
| C1=CC(=NC=C1C#N)Cl | |
| 138.554 | |
| 138.55 | |
| Crystalline Powder |
| 33252-28-7 | |
| 107°C | |
| MFCD00084941 | |
| 3439 | |
| ORIQLMBUPMABDV-UHFFFAOYSA-N | |
| 6-chloropyridine-3-carbonitrile | |
| 5152094 | |
| ≥97.0% (GC,N) |
Chemical Identifiers
| 33252-28-7 | |
| 138.554 | |
| ORIQLMBUPMABDV-UHFFFAOYSA-N | |
| 5152094 | |
| C1=CC(=NC=C1C#N)Cl |
| C6H3ClN2 | |
| MFCD00084941 | |
| 2-chloro-5-cyanopyridine, 6-chloronicotinonitrile, 2-chloropyridine-5-carbonitrile, 6-chloro-3-pyridinecarbonitrile, 6-chloronicotinitrile, 6-chloronicotinontrile, 6-chloro-nicotinonitrile, 2-chloro-5-cyano-pyridine, 3-pyridinecarbonitrile, 6-chloro, 6-chloro nicotinonitrile | |
| 6-chloropyridine-3-carbonitrile |
Safety and Handling
TSCA : No