accessibility menu, dialog, popup

UserName

ClNN

CAS RN 33252-28-7

IUPAC Name: 6-chloropyridine-3-carbonitrile
Synonyms: 6-chloropyridine-3-carbonitrile
Molecular Weight (g/mol): 138.55
Molecular Formula: C6H3ClN2
InChi Key: ORIQLMBUPMABDV-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=N1)C#N
Boiling Point 
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)