missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(chloromethyl)pyridine 98.0+%, TCI America™

Catalog No. c162825g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C162825G 25 g
C16285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C162825G Supplier TCI America Supplier No. C162825G
Only null left

Chemical Identifiers

CAS 70258-18-3
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00125366
InChI Key SKCNYHLTRZIINA-UHFFFAOYSA-N
Synonym 2-chloro-5-chloromethyl pyridine, pyridine, 2-chloro-5-chloromethyl, 2-chloro-5-chloromethyl-pyridine, pubchem9210, acmc-1bl9y, ksc377c3b, 2-chloro-5chloromethylpyridine, 2-chloro-5-chloromethypyridine, 5-chloromethyl-2-chloropyridine, 6-chloro-3-chloromethylpyridine
PubChem CID 155479
IUPAC Name 2-chloro-5-(chloromethyl)pyridine
SMILES C1=CC(=NC=C1CCl)Cl

Specifications

Melting Point 39°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 162.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-(chloromethyl)pyridine