Learn More
2-Chloro-5-(chloromethyl)pyridine, 96%
CAS: 70258-18-3 | C6H5Cl2N | 162.013 g/mol
$85.97 - $210.07
Chemical Identifiers
| CAS | 70258-18-3 |
|---|---|
| Molecular Formula | C6H5Cl2N |
| Molecular Weight (g/mol) | 162.013 |
| MDL Number | MFCD00125366 |
| InChI Key | SKCNYHLTRZIINA-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-chloromethyl pyridine, pyridine, 2-chloro-5-chloromethyl, 2-chloro-5-chloromethyl-pyridine, pubchem9210, acmc-1bl9y, ksc377c3b, 2-chloro-5chloromethylpyridine, 2-chloro-5-chloromethypyridine, 5-chloromethyl-2-chloropyridine, 6-chloro-3-chloromethylpyridine |
| PubChem CID | 155479 |
| IUPAC Name | 2-chloro-5-(chloromethyl)pyridine |
| SMILES | C1=CC(=NC=C1CCl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1928406
|
Thermo Scientific Chemicals
L1928406 |
5 g |
N/A
|
|
|||||
|
AAL1928414
|
Thermo Scientific Chemicals
L1928414 |
25 g |
N/A
|
|
|||||
Description
2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-5-(chloromethyl)pyridine is used for the synthesis of various pharmaceutical compounds. It can also be used for the synthesis of new neonicotinoid compounds.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 70258-18-3 | |
| 162.013 | |
| SKCNYHLTRZIINA-UHFFFAOYSA-N | |
| 155479 | |
| C1=CC(=NC=C1CCl)Cl |
| C6H5Cl2N | |
| MFCD00125366 | |
| 2-chloro-5-chloromethyl pyridine, pyridine, 2-chloro-5-chloromethyl, 2-chloro-5-chloromethyl-pyridine, pubchem9210, acmc-1bl9y, ksc377c3b, 2-chloro-5chloromethylpyridine, 2-chloro-5-chloromethypyridine, 5-chloromethyl-2-chloropyridine, 6-chloro-3-chloromethylpyridine | |
| 2-chloro-5-(chloromethyl)pyridine |
Specifications
| 70258-18-3 | |
| >110°C (230°F) | |
| MFCD00125366 | |
| UN3261 | |
| 2-chloro-5-chloromethyl pyridine, pyridine, 2-chloro-5-chloromethyl, 2-chloro-5-chloromethyl-pyridine, pubchem9210, acmc-1bl9y, ksc377c3b, 2-chloro-5chloromethylpyridine, 2-chloro-5-chloromethypyridine, 5-chloromethyl-2-chloropyridine, 6-chloro-3-chloromethylpyridine | |
| SKCNYHLTRZIINA-UHFFFAOYSA-N | |
| 2-chloro-5-(chloromethyl)pyridine | |
| 155479 | |
| 96% |
| 37°C to 42°C | |
| C6H5Cl2N | |
| 5 g | |
| 1635690 | |
| Insoluble in water. | |
| C1=CC(=NC=C1CCl)Cl | |
| 162.013 | |
| 162.02 | |
| 2-Chloro-5-(chloromethyl)pyridine |
Safety and Handling
GHS H Statement
H314-H318-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H334-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only