missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)pyridine, 98+%

CAS: 81565-18-6 | C6H3ClF3N | 181.54 g/mol

$139.63 - $488.99

Chemical Identifiers

CAS 81565-18-6
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042224
InChI Key GBNPVXZNWBWNEN-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine
PubChem CID 144958
IUPAC Name 2-chloro-4-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=NC(Cl)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1071303
View Documents
Thermo Scientific Chemicals
L1071303
1 g
Each for $139.63
Only null left
 
AAL1071306
View Documents
Thermo Scientific Chemicals
L1071306
5 g
Each for $488.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

81565-18-6
181.54
GBNPVXZNWBWNEN-UHFFFAOYSA-N
144958
FC(F)(F)C1=CC=NC(Cl)=C1
C6H3ClF3N
MFCD00042224
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine
2-chloro-4-(trifluoromethyl)pyridine

Specifications

81565-18-6
145°C
C6H3ClF3N
MFCD00042224
UN1992
2-chloro-4-trifluoromethyl pyridine, pyridine, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoro methylpyridine, 2-chloro-4-trifluoromethyl-pyridine, pubchem2204, acmc-209plt, ksc449e5r, 2-chloro-4-trifluromethylpyridine, 2-chloro-4-trifluoro-methylpyridine
FC(F)(F)C1=CC=NC(Cl)=C1
181.54
181.55
2-Chloro-4-(trifluoromethyl)pyridine
1.411
40°C (104°F)
1.449
1 g
4246276
GBNPVXZNWBWNEN-UHFFFAOYSA-N
2-chloro-4-(trifluoromethyl)pyridine
144958
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H302-H312-H315-H319-H335
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only